siRNA Details
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001552.4 | NP_001001552.3 | 2 | 182 | ||
NM_001199050.1 | NP_001185979.1 | 2 | 182 | |||
NM_001199051.1 | NP_001185980.1 | 2 | 182 | |||
NM_001199052.1 | NP_001185981.1 | 2 | 182 | |||
NR_037583.1 | 2 | 182 | ||||
XM_011510160.2 | XP_011508462.1 | 2 | 186 | |||
XM_011510161.2 | XP_011508463.1 | 2 | 174 | |||
XM_011510162.2 | XP_011508464.1 | 2 | 184 | |||
XM_011510163.2 | XP_011508465.1 | 2 | 189 | |||
XM_011510164.2 | XP_011508466.1 | 2 | 206 | |||
XM_011510165.2 | XP_011508467.1 | 2 | 202 | |||
GenBank mRNA | AB084764.1 | - | 2 | 149 | ||
AB084765.1 | - | 2 | 149 | |||
AB085610.1 | - | 2 | 149 | |||
AB096677.1 | - | 2 | 149 | |||
AB096678.1 | - | 2 | 149 | |||
AB096679.1 | - | 2 | 149 | |||
AB096680.1 | - | 2 | 149 | |||
AB096681.1 | - | 2 | 149 | |||
AB096682.1 | - | 2 | 149 | |||
AK026628.1 | - | 2 | 149 | |||
AK302392.1 | - | 2 | 184 | |||
BC036636.2 | - | 2 | 152 | 5296437 | ||
DC399934.1 | - | 2 | 184 |
