siRNA Details
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 3 | 327 | ||
NM_178190.2 | NP_835497.1 | 3 | 403 | |||
NM_178191.2 | NP_835498.1 | 2 | 1711 | |||
GenBank mRNA | AB029042.1 | - | 3 | 255 | ||
AF114836.1 | - | 413 | ||||
AK000934.1 | - | 2 | 1648 | |||
AK316600.1 | - | 3 | 314 | |||
AL050386.1 | - | 2 | 1641 | |||
AL583540.3 | - | 3 | 326 | |||
BC004955.1 | - | 3 | 210 | 3625379 | ||
BC009677.1 | - | 3 | 285 | 3877506 | ||
BG036978.1 | - | 3 | 375 | |||
BG337531.1 | - | 3 | 283 | |||
BP271519.1 | - | 3 | 323 | |||
BT009849.1 | - | 3 | 234 | |||
CR457097.1 | - | 3 | 234 |
