siRNA Details
Gene Symbol: CPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127441.1 | NP_001120913.1 | 13 | 1709 | ||
NM_001318223.1 | NP_001305152.1 | 11 | 1598 | |||
NM_080385.4 | NP_525124.3 | 12 | 1656 | |||
XM_005250710.1 | XP_005250767.1 | 12 | 1637 | |||
XM_005250712.1 | XP_005250769.1 | 11 | 1551 | |||
XM_011516698.1 | XP_011515000.1 | 11 | 2274 | |||
XM_011516701.1 | XP_011515003.1 | 11 | 2191 | |||
XM_011516702.1 | XP_011515004.1 | 11 | 2177 | |||
XM_011516703.1 | XP_011515005.1 | 11 | 2170 | |||
GenBank mRNA | AF384667.1 | - | 10 | 1089 | ||
AK093288.1 | - | 10 | 1645 | |||
AK315256.1 | - | 11 | 1572 | |||
AY155229.1 | - | 12 | 1609 | |||
AY155230.1 | - | 13 | 1679 | |||
BC039362.1 | - | 13 | 1683 | 5270617 | ||
BC042996.1 | - | 11 | 2225 | 5296768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
