siRNA Details
Gene Symbol: TTYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032646.5 | NP_116035.5 | 11, 12 | 1295 | ||
NM_052869.1 | NP_443101.1 | 5, 6 | 1105 | |||
XM_005257824.3 | XP_005257881.1 | 11, 12 | 1223 | |||
GenBank mRNA | AF319952.1 | - | 11, 12 | 1274 | ||
AK126955.1 | - | 5, 6 | 1513 | |||
AK296511.1 | - | 11, 12 | 1218 | |||
BC005168.1 | - | 11, 12 | 1270 | |||
BC107602.1 | - | 11, 12 | 1291 | 40033791 | ||
BX647912.1 | - | 5, 6 | 1105 |
