siRNA Details
Gene Symbol: ZNF101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 1273 | ||
NM_033204.3 | NP_149981.2 | 4 | 1401 | |||
GenBank mRNA | AK097169.1 | - | 4 | 1393 | ||
AY149174.1 | - | 4 | 1335 | |||
BC011502.1 | - | 1 | 1037 | |||
BC119680.1 | - | 5 | 1437 | 40116294 |
