siRNA Details
Gene Symbol: SYTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292588.1 | - | 13 | 1533 | ||
AK313086.1 | - | 12 | 1418 | |||
AL832596.1 | - | 10 | 2832 | |||
BC014913.1 | - | 14 | 1588 | 3908307 | ||
BX537410.1 | - | 13 | 1531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
