siRNA Details
Gene Symbol: SYTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129896.2 | NP_001123368.1 | 9, 10 | 1116 | ||
NM_001174068.1 | NP_001167539.1 | 8, 9 | 995 | |||
NM_080737.2 | NP_542775.2 | 10, 11 | 1164 | |||
XM_005262229.2 | XP_005262286.1 | 8, 9 | 996 | |||
XM_011531068.2 | XP_011529370.1 | 10, 11 | 1193 | |||
XM_017029966.1 | XP_016885455.1 | 10, 11 | 1193 | |||
XM_017029967.1 | XP_016885456.1 | 9, 10 | 1085 | |||
XM_017029968.1 | XP_016885457.1 | 8, 9 | 996 | |||
XM_017029969.1 | XP_016885458.1 | 8, 9 | 970 | |||
XM_017029970.1 | XP_016885459.1 | 8, 9 | 918 | |||
XM_017029971.1 | XP_016885460.1 | 9, 10 | 1096 | |||
GenBank mRNA | AF302493.1 | - | 7, 8 | 881 | ||
AK292588.1 | - | 9, 10 | 1085 | |||
AK313086.1 | - | 8, 9 | 970 | |||
AL832596.1 | - | 8, 9 | 995 | |||
BC014913.1 | - | 10, 11 | 1140 | 3908307 | ||
BX537410.1 | - | 9, 10 | 1083 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
