siRNA Details
Gene Symbol: SNX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052870.2 | NP_443102.2 | 1 | 1994 | ||
GenBank mRNA | AF395536.1 | - | 1 | 1804 | ||
BC117218.1 | - | 1 | 1892 | 40125769 | ||
BC117220.1 | - | 1 | 1892 | 40125771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
