siRNA Details
Gene Symbol: IFT43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052873.2 | NP_443105.2 | 4 | 291 | ||
GenBank mRNA | AK056735.1 | - | 6 | 344 | ||
BC010436.1 | - | 4 | 262 | 3608220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
