siRNA Details
Gene Symbol: HOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138413.3 | NP_612422.2 | 3, 4 | 825 | ||
GenBank mRNA | AK289440.1 | - | 3, 4 | 492 | ||
BC045550.1 | - | 3, 4 | 809 | 5274464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
