siRNA Details
Gene Symbol: PGLYRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722633.3 | XP_006722696.1 | 4 | 1911 | ||
GenBank mRNA | AK055882.1 | - | 4 | 1911 | ||
BC136551.1 | - | 4 | 1803 | 9020541 | ||
BC136552.1 | - | 4 | 1803 | 9020542 | ||
BC144238.1 | - | 4 | 1803 | 9052758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
