siRNA Details
Gene Symbol: TUBGCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102610.1 | NP_001096080.1 | 13 | 1676 | ||
NM_052903.4 | NP_443135.3 | 13 | 1676 | |||
XM_005272540.2 | XP_005272597.1 | 13 | 1583 | |||
XM_011543868.1 | XP_011542170.1 | 13 | 1583 | |||
XM_011543869.2 | XP_011542171.1 | 13 | 1580 | |||
XM_011543870.1 | XP_011542172.1 | 13 | 1583 | |||
XM_017021890.1 | XP_016877379.1 | 13 | 1583 | |||
XM_017021891.1 | XP_016877380.1 | 13 | 1580 | |||
XM_017021892.1 | XP_016877381.1 | 13 | 1580 | |||
XM_017021893.1 | XP_016877382.1 | 13 | 1583 | |||
XM_017021894.1 | XP_016877383.1 | 10 | 1231 | |||
XM_017021895.1 | XP_016877384.1 | 13 | 1600 | |||
XM_017021896.1 | XP_016877385.1 | 5 | 715 | |||
XM_017021897.1 | XP_016877386.1 | 4 | 475 | |||
XM_017021898.1 | XP_016877387.1 | 8 | 1036 | |||
XM_017021899.1 | XP_016877388.1 | 13 | 1621 | |||
XM_017021900.1 | XP_016877389.1 | 12 | 1496 | |||
XM_017021901.1 | XP_016877390.1 | 4 | 1545 | |||
GenBank mRNA | AB067486.2 | - | 13 | 1590 | ||
AF272884.1 | - | 13 | 1600 | |||
AK023934.1 | - | 4 | 1728 | |||
AK056416.1 | - | 13 | 1575 | |||
AK289859.1 | - | 13 | 1575 | |||
BC046182.1 | - | 2 | 101 | |||
BC071560.1 | - | 13 | 1572 | 5297149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
