siRNA Details
Gene Symbol: RNF157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052916.2 | NP_443148.1 | 3 | 334 | ||
XM_005257007.4 | XP_005257064.1 | 3 | 393 | |||
XM_011524273.2 | XP_011522575.1 | 3 | 393 | |||
XM_017024117.1 | XP_016879606.1 | 3 | 393 | |||
XM_017024118.1 | XP_016879607.1 | 3 | 393 | |||
XM_017024119.1 | XP_016879608.1 | 3 | 393 | |||
XM_017024120.1 | XP_016879609.1 | 3 | 190 | |||
GenBank mRNA | AB067504.2 | - | 3 | 334 | ||
AK091467.1 | - | 3 | 217 |
