siRNA Details
Gene Symbol: KCTD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138444.3 | NP_612453.1 | 1 | 1908 | ||
GenBank mRNA | AF359381.1 | - | 1 | 1893 | ||
BC013764.1 | - | 1 | 1738 | 4521096 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCTD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138444.3 | NP_612453.1 | 1 | 1908 | ||
GenBank mRNA | AF359381.1 | - | 1 | 1893 | ||
BC013764.1 | - | 1 | 1738 | 4521096 |