siRNA Details
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300764.1 | NP_001287693.1 | 4 | 497 | ||
NM_052965.3 | NP_443197.1 | 4 | 497 | |||
NR_023349.2 | 5 | 614 | ||||
NR_125335.1 | 5 | 577 | ||||
XM_006711148.1 | XP_006711211.1 | 4 | 446 | |||
GenBank mRNA | AF288394.1 | - | 4 | 472 | ||
AK098196.1 | - | 4 | 395 | |||
AK307241.1 | - | 5 | 505 | |||
BC022030.1 | - | 4 | 442 | 4824025 | ||
BP326115.1 | - | 4 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
