siRNA Details
Gene Symbol: FAM129A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052966.3 | NP_443198.1 | 13 | 1833 | ||
XM_011509140.2 | XP_011507442.1 | 14 | 2708 | |||
XM_011509141.2 | XP_011507443.1 | 13 | 2597 | |||
GenBank mRNA | AB050477.1 | - | 13 | 1759 | ||
AF288391.1 | - | 13 | 1797 | |||
AK022527.1 | - | 13 | 1671 | |||
AK074139.1 | - | 1 | 41 | |||
AK095547.1 | - | 8 | 1077 | |||
AY550972.1 | - | 13 | 1672 | |||
BC030531.1 | - | 13 | 1689 | 5212338 |
