siRNA Details
Gene Symbol: MRPL54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172251.2 | NP_758455.1 | 3 | 521 | ||
GenBank mRNA | BC065273.1 | - | 3 | 514 | 6165136 | |
BF346319.1 | - | 3 | 514 | |||
BM792364.1 | - | 3 | 481 | |||
BQ061807.1 | - | 3 | 509 |
