siRNA Details
Gene Symbol: TRIM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136486.1 | NP_001129958.1 | 1, 2 | 408 | ||
XM_011542585.2 | XP_011540887.1 | 2, 3 | 497 |
Gene Symbol: TRIM64B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164397.1 | NP_001157869.1 | 1, 2 | 408 | ||
XM_011542954.2 | XP_011541256.1 | 2, 3 | 497 | |||
GenBank mRNA | JF968449.1 | - | 1, 2 | 393 |
Gene Symbol: TRIM64DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968450.1 | - | 1, 2 | 400 |
Gene Symbol: TRIM64EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968451.1 | - | 1, 2 | 401 |
