siRNA Details
Gene Symbol: CCDC168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146197.1 | NP_001139669.1 | 4 | 15740 | ||
XM_011521106.1 | XP_011519408.1 | 5 | 15613 | |||
GenBank mRNA | AK097495.1 | - | 1 | 1723 |
