siRNA Details
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144609.2 | NP_653210.2 | 4, 5 | 539 | ||
GenBank mRNA | AK056357.1 | - | 4, 5 | 504 | ||
AK098464.1 | - | 12 | ||||
BC023298.1 | - | 4, 5 | 500 | |||
BC047776.2 | - | 4, 5 | 514 | 6023964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
