siRNA Details
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145046.4 | NP_659483.2 | 5 | 587 | ||
GenBank mRNA | AB576176.1 | - | 5 | 524 | ||
AK058084.1 | - | 5 | 579 | |||
BC014595.2 | - | 5 | 587 | 4822010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
