siRNA Details
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB576176.1 | - | 3 | 378 | ||
AK058084.1 | - | 3 | 433 | |||
BC014595.2 | - | 3 | 441 | 4822010 | ||
DB459545.1 | - | 3 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
