siRNA Details
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002912.4 | NP_001002912.4 | 8 | 1089 | ||
XM_017000274.1 | XP_016855763.1 | 8 | 1089 | |||
XM_017000275.1 | XP_016855764.1 | 8 | 879 | |||
GenBank mRNA | AK127470.1 | - | 8 | 1089 | ||
BC073916.1 | - | 3 | 457 | |||
BI832067.1 | - | 3 | 448 |
