siRNA Details
Gene Symbol: ZNF648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009992.1 | NP_001009992.1 | 2 | 1283 | ||
XM_017000301.1 | XP_016855790.1 | 4 | 4710 | |||
XM_017000302.1 | XP_016855791.1 | 6 | 5216 | |||
GenBank mRNA | AK128654.1 | - | 2 | 1283 | ||
BC137223.1 | - | 2 | 1186 | 9021220 | ||
BC137224.1 | - | 2 | 1186 | 9021221 |
