siRNA Details
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300831.1 | NP_001287760.1 | 2 | 187 | ||
NM_181643.5 | NP_857594.2 | 2 | 187 | |||
NR_125338.1 | 2 | 187 | ||||
XM_005270472.1 | XP_005270529.1 | 2 | 296 | |||
XM_017000322.1 | XP_016855811.1 | 2 | 271 | |||
GenBank mRNA | AK074433.1 | - | 2 | 122 | ||
AK303897.1 | - | 2 | 148 | |||
AK310801.1 | - | 2 | 138 | |||
BC029237.1 | - | 1 | 76 | |||
BC050319.1 | - | 2 | 151 | 4837146 | ||
BC068603.1 | - | 2 | 111 | |||
BC093891.1 | - | 2 | 174 | 7939736 | ||
BC101501.1 | - | 2 | 175 | 8069007 | ||
BF674731.1 | - | 2 | 189 | |||
HM237139.1 | - | 2 | 174 | |||
HM237140.1 | - | 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
