siRNA Details
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300831.1 | NP_001287760.1 | 3 | 354 | ||
NM_181643.5 | NP_857594.2 | 4 | 453 | |||
XM_005270472.1 | XP_005270529.1 | 4 | 562 | |||
XM_017000322.1 | XP_016855811.1 | 3 | 438 | |||
GenBank mRNA | AK074433.1 | - | 4 | 388 | ||
AK303897.1 | - | 3 | 315 | |||
BC029237.1 | - | 3 | 342 | |||
BC068603.1 | - | 4 | 377 | |||
BC093891.1 | - | 4 | 440 | 7939736 | ||
BC101501.1 | - | 4 | 441 | 8069007 | ||
BF674731.1 | - | 4 | 459 | |||
HM237139.1 | - | 4 | 440 | |||
HM237140.1 | - | 4 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
