siRNA Details
Gene Symbol: TSHZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193421.1 | NP_001180350.1 | 2 | 2252 | ||
NM_173485.5 | NP_775756.3 | 2 | 2785 | |||
XM_017027640.1 | XP_016883129.1 | 2 | 2883 | |||
XM_017027641.1 | XP_016883130.1 | 2 | 2883 | |||
GenBank mRNA | AK091206.1 | - | 2 | 1978 | ||
AK302570.1 | - | 2 | 2252 | |||
BX640770.1 | - | 2 | 2785 | |||
BX648148.1 | - | 1 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
