siRNA Details
Gene Symbol: TRABD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277053.1 | NP_001263982.1 | 2, 3 | 882 | ||
NM_001307978.1 | NP_001294907.1 | 2, 3 | 882 | |||
XM_011532504.1 | XP_011530806.1 | 2, 3 | 682 | |||
GenBank mRNA | AK296608.1 | - | 2, 3 | 719 | ||
AK309615.1 | - | 2, 3 | 721 | |||
JQ653415.1 | - | 2, 3 | 676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
