siRNA Details
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194282.3 | NP_919258.2 | 2, 3 | 1068 | ||
XM_005262750.4 | XP_005262807.1 | 2, 3 | 877 | |||
XM_006714081.3 | XP_006714144.1 | 2, 3 | 803 | |||
XM_017007729.1 | XP_016863218.1 | 2, 3 | 1010 | |||
GenBank mRNA | AK294256.1 | - | 2, 3 | 997 | ||
BX640966.1 | - | 2, 3 | 1063 | |||
CX871759.1 | - | 1, 2 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
