siRNA Details
Gene Symbol: C5orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001089584.2 | NP_001083053.1 | 2 | 720 | ||
XM_011513963.2 | XP_011512265.1 | 2 | 271 | |||
GenBank mRNA | BC053548.1 | - | 2 | 373 | ||
BC062569.1 | - | 2 | 720 | |||
BC104204.2 | - | 2 | 381 | |||
BC104205.2 | - | 2 | 381 | 40031408 | ||
BC115361.1 | - | 2 | 381 | 40031409 |
