siRNA Details
Gene Symbol: C5orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001089584.2 | NP_001083053.1 | 3 | 1605 | ||
XM_011513963.2 | XP_011512265.1 | 3 | 1156 | |||
GenBank mRNA | BC053548.1 | - | 3 | 1258 | ||
BC062569.1 | - | 3 | 1605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
