siRNA Details
Gene Symbol: OR6B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005281.1 | NP_001005281.1 | 1 | 584 | ||
GenBank mRNA | BC137042.1 | - | 1 | 652 | 9021038 | |
BC137047.1 | - | 1 | 652 | 9021044 |
