siRNA Details
Gene Symbol: C9orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128618.1 | NP_001122090.1 | 1 | 92 | ||
XM_011518225.2 | XP_011516527.1 | 1 | 86 | |||
GenBank mRNA | BC036255.1 | - | 1 | 92 | ||
BC044620.1 | - | 1 | 70 | |||
BC053956.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
