siRNA Details
Gene Symbol: OTUD6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207320.2 | NP_997203.1 | 1 | 149 | ||
GenBank mRNA | AK098697.1 | - | 1 | 149 | ||
BC137355.1 | - | 1 | 145 | 9021353 | ||
BC137356.1 | - | 1 | 145 | 9021354 |
