siRNA Details
Gene Symbol: MROH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152503.5 | NP_689716.4 | 5, 6 | 859 | ||
NM_213631.2 | NP_998796.1 | 5, 6 | 859 | |||
NM_213632.2 | NP_998797.2 | 5, 6 | 859 | |||
GenBank mRNA | AK093432.1 | - | 5, 6 | 859 | ||
BC030006.1 | - | 5, 6 | 836 | 4822338 | ||
BC057767.1 | - | 5, 6 | 842 | 5272440 |
