siRNA Details
Gene Symbol: MROH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152503.5 | NP_689716.4 | 9, 10 | 1348 | ||
NM_213631.2 | NP_998796.1 | 9, 10 | 1348 | |||
NM_213632.2 | NP_998797.2 | 8, 9 | 1243 | |||
GenBank mRNA | AK093432.1 | - | 8, 9 | 1243 | ||
AL833864.1 | - | 2, 3 | 245 | |||
BC030006.1 | - | 9, 10 | 1325 | 4822338 | ||
BC057767.1 | - | 9, 10 | 1331 | 5272440 | ||
BX098572.1 | - | 1, 2 | 147 |
