siRNA Details
Gene Symbol: ISM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080826.1 | NP_543016.1 | 2, 3 | 383 | ||
XM_017027680.1 | XP_016883169.1 | 2, 3 | 383 | |||
GenBank mRNA | CR982468.1 | - | 1, 2 | 164 | ||
DB169194.1 | - | 2, 3 | 385 |
