siRNA Details
Gene Symbol: HOXB-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033202.2 | 2 | 226 | |||
NR_033203.1 | 2 | 129 | ||||
NR_033205.1 | 2 | 222 | ||||
NR_110331.1 | 2 | 226 | ||||
GenBank mRNA | AA299267.1 | - | 2 | 166 | ||
BG196429.1 | - | 1 | 31 | |||
BG203124.1 | - | 2 | 257 | |||
BG203609.1 | - | 2 | 252 | |||
CB046509.1 | - | 2 | 138 | |||
CF127739.1 | - | 2 | 204 | |||
DA741273.1 | - | 2 | 231 |
