siRNA Details
Gene Symbol: PSMA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025096.1 | NP_001020267.1 | 3 | 451 | ||
NM_001025097.1 | NP_001020268.1 | 3 | 337 | |||
NM_001308188.1 | NP_001295117.1 | 4 | 521 | |||
NM_144662.2 | NP_653263.2 | 3 | 469 | |||
XM_011525818.1 | XP_011524120.1 | 4 | 404 | |||
GenBank mRNA | BC021713.1 | - | 4 | 461 | 4826048 | |
BC025389.1 | - | 3 | 447 | |||
BC025393.1 | - | 3 | 325 | |||
BC028371.1 | - | 3 | 325 | 4828792 | ||
BC028686.1 | - | 3 | 325 | 4828931 | ||
BC042820.1 | - | 3 | 451 | 5268996 | ||
BC047355.1 | - | 3 | 443 | |||
BI461560.1 | - | 3 | 456 |
