siRNA Details
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153705.4 | NP_714916.3 | 6 | 1307 | ||
XM_011542621.2 | XP_011540923.1 | 6 | 1280 | |||
XM_011542623.2 | XP_011540925.1 | 4 | 860 | |||
GenBank mRNA | AF533708.1 | - | 6 | 1266 | ||
AK098206.1 | - | 2 | 272 | |||
AK125519.1 | - | 5 | 1089 | |||
AK313534.1 | - | 6 | 1169 | |||
AY358616.1 | - | 5 | 1376 | |||
BC036526.2 | - | 6 | 1261 | 5266665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
