siRNA Details
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 15 | 1209 | ||
XM_011537951.2 | XP_011536253.1 | 15 | 4184 | |||
XM_017018851.1 | XP_016874340.1 | 16 | 2182 | |||
XM_017018852.1 | XP_016874341.1 | 15 | 1155 | |||
GenBank mRNA | AK074223.1 | - | 1 | 71 | ||
AK094998.1 | - | 3 | 783 | |||
AK098593.1 | - | 15 | 1140 | |||
AK307908.1 | - | 15 | 1180 | |||
AY177785.1 | - | 15 | 1125 | |||
BC046366.1 | - | 3 | 526 | |||
BC110577.1 | - | 15 | 1200 | 40007743 | ||
BC110578.1 | - | 15 | 1200 | 40007744 | ||
BX538055.1 | - | 15 | 1223 |
