siRNA Details
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 1466 | ||
NM_001177997.1 | NP_001171468.1 | 3 | 1359 | |||
NM_144671.4 | NP_653272.2 | 3 | 1378 | |||
GenBank mRNA | AK056918.1 | - | 3 | 764 | ||
AK093052.1 | - | 2 | 1082 | |||
AK299344.1 | - | 3 | 1328 | |||
BC034809.1 | - | 3 | 1279 | 4943538 | ||
CX785083.1 | - | 1 | 243 |
