siRNA Details
Gene Symbol: C19orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193623.1 | NP_001180552.1 | 2 | 899 | ||
NR_126530.1 | 2 | 1028 | ||||
GenBank mRNA | BC043532.1 | - | 2 | 899 | ||
BC085012.1 | - | 2 | 872 |
