siRNA Details
Gene Symbol: EFNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004429.4 | NP_004420.1 | 2 | 928 | ||
GenBank mRNA | BC016649.1 | - | 2 | 928 | 3867060 | |
BC052979.1 | - | 2 | 947 | 6164440 | ||
L37361.1 | - | 2 | 674 | |||
U09303.1 | - | 2 | 849 | |||
U09304.1 | - | 2 | 455 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: Z48781, AK034996, BC006797, U12983, BC021656, and others.
Gene Ontology Categories:
