siRNA Details
Gene Symbol: ZNF599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007248.2 | NP_001007249.1 | 4 | 1448 | ||
XM_017026369.1 | XP_016881858.1 | 4 | 1523 | |||
GenBank mRNA | AK055225.1 | - | 4 | 1434 | ||
BC044615.1 | - | 5 | 1564 | 5266705 |
