siRNA Details
Gene Symbol: ZNF555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172775.1 | NP_001166246.1 | 4 | 1862 | ||
NM_152791.4 | NP_690004.4 | 4 | 1865 | |||
XM_011527716.2 | XP_011526018.1 | 4 | 1865 | |||
XM_017026375.1 | XP_016881864.1 | 4 | 1865 | |||
GenBank mRNA | AK056659.1 | - | 1 | 1163 | ||
AK093163.1 | - | 6 | 1598 | |||
AK292954.1 | - | 4 | 1810 | |||
AL832140.1 | - | 4 | 1826 | |||
BC022022.2 | - | 4 | 1816 | 4822197 | ||
BX647423.1 | - | 4 | 1823 |
