siRNA Details
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288579.1 | NP_001275508.1 | 11, 12 | 1455 | ||
NM_001288580.1 | NP_001275509.1 | 11, 12 | 1161 | |||
NM_144695.3 | NP_653296.2 | 12, 13 | 1551 | |||
XM_005273065.2 | XP_005273122.2 | 12, 13 | 1187 | |||
XM_005273069.4 | XP_005273126.1 | 11, 12 | 1423 | |||
XM_006711173.3 | XP_006711236.1 | 13, 14 | 1620 | |||
XM_011509212.2 | XP_011507514.1 | 11, 12 | 1514 | |||
XM_011509213.2 | XP_011507515.1 | 11, 12 | 1495 | |||
XM_011509214.2 | XP_011507516.1 | 13, 14 | 1420 | |||
XM_017000374.1 | XP_016855863.1 | 14, 15 | 1745 | |||
XM_017000375.1 | XP_016855864.1 | 10, 11 | 1400 | |||
XM_017000376.1 | XP_016855865.1 | 12, 13 | 1548 | |||
GenBank mRNA | AB276123.1 | - | 11, 12 | 1145 | ||
AK056983.1 | - | 12, 13 | 1540 | |||
AK293974.1 | - | 11, 12 | 1087 | |||
AK315930.1 | - | 11, 12 | 1091 | |||
BC113635.1 | - | 12, 13 | 1288 | 8322687 | ||
BC113637.1 | - | 12, 13 | 1288 | 8322689 | ||
BC143811.1 | - | 11, 12 | 1192 | 9052326 |
