siRNA Details
Gene Symbol: KNCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097611.1 | NP_001091080.1 | 3 | 1258 | ||
NM_001322255.1 | NP_001309184.1 | 4 | 1769 | |||
GenBank mRNA | AK056573.1 | - | 3 | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
