siRNA Details
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148960.2 | NP_683763.2 | 4, 5 | 819 | ||
GenBank mRNA | AK096063.1 | - | 4, 5 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
