siRNA Details
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123395.1 | NP_001116867.1 | 4 | 2325 | ||
NM_001185117.1 | NP_001172046.1 | 3 | 2240 | |||
NM_148960.2 | NP_683763.2 | 5 | 1582 | |||
GenBank mRNA | AK056118.1 | - | 1 | 781 | ||
AK096063.1 | - | 5 | 1582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
